Tisk…

Curcumin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

C1386-10G C1386-50G C1386-5G
SIALC1386-10GEA 2890 CZK
SIALC1386-10G SIALC1386-50G SIALC1386-5G
Curcumin, Sigma-Aldrich®
Curcumin
Vzorec: C₂₁H₂₀O₆
MDL Number: MFCD00008365
CAS číslo: 458-37-7
EINECS: 207-280-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR