Tisk…

2-Amino-4-chloro-6-methylpyrimidine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Chloro-6-methylpyrimidin-2-amine

M-Clarity™ quality level = MQ100

122882-25G 122882-5G
SIAL122882-25GEA 4750 CZK
SIAL122882-25G SIAL122882-5G
2-Amino-4-chloro-6-methylpyrimidine, Sigma-Aldrich®
2-Amino-4-chloro-6-methylpyrimidine
Vzorec: C₅H₆ClN₃
MW: 143,58 g/mol
MDL Number: MFCD00006091
CAS číslo: 5600-21-5
EINECS: 227-018-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR