4-Chloro-2,6-diaminopyrimidine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,4-Diamino-6-chloropyrimidine, 6-Chloropyrimidine-2,4-diamine
M-Clarity™ quality level = MQ100
C33204-25G
C33204-5G
SIALC33204-25GEA
4390
CZK
SIALC33204-25G
SIALC33204-5G
4-Chloro-2,6-diaminopyrimidine, Sigma-Aldrich®
4-Chloro-2,6-diaminopyrimidine
Vzorec:
C₄H₅ClN₄ MW: 144,56 g/mol |
MDL Number:
MFCD00006097 CAS číslo: 156-83-2 EINECS: 205-863-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...