3-Amino-1-methyl-1H-pyrazole 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1-Methyl-1H-pyrazol-3-amine, 3-Amino-1-methylpyrazole, 1-Methyl-1H-pyrazol-3-ylamine, 3-Amino-1-methyl-1H-pyrazole
H30935.03
H30935.06
H30935.03EA
652
CZK
H30935.03
H30935.06
3-Amino-1-methyl-1H-pyrazole 97%
3-Amino-1-methyl-1H-pyrazole
Purity: 97%
CAS No.: 1904-31-0
Molecular Formula: C4H7N3
Formula Weight: 97.12
Boiling Point: 90-92°/1mm
UN No.: 2735
MDL No.: MFCD00466340
Vzorec:
C₄H₇N₃ Bod varu: 90…92 °C (1 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00466340 CAS číslo: 1904-31-0 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...