Tisk…

1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(1-Ethyl-1H-pyrazol-4yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Ethyl-1H-pyrazole-4-boronic acid Pinacol ester, 1-Ethyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

M-Clarity™ quality level = MQ100

706205-1G 706205-250MG
SIAL706205-1GEA 8470 CZK
SIAL706205-1G SIAL706205-250MG
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester, Sigma-Aldrich®
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester
Vzorec: C₁₁H₁₉BN₂O₂
MW: 222.0918 g/mol
MDL Number: MFCD09864188
CAS číslo: 847818-70-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR