2,6-Dimethyl-p-benzoquinone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
D149705-1G
D149705-5G
SIALD149705-1GEA
1720
CZK
SIALD149705-1G
SIALD149705-5G
2,6-Dimethyl-p-benzoquinone, Sigma-Aldrich®
2,6-Dimethyl-p-benzoquinone
Vzorec:
C₈H₈O₂ |
MDL Number:
MFCD00001605 CAS číslo: 527-61-7 EINECS: 208-420-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...