Tisk…

2,6-Dimethoxy-p-benzoquinone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,6-Dimethoxy-1,4-Benzoquinone

M-Clarity™ quality level = MQ100

428566-25G 428566-5G
SIAL428566-25GEA 7100 CZK
SIAL428566-25G SIAL428566-5G
2,6-Dimethoxy-p-benzoquinone, Sigma-Aldrich®
2,6-Dimethoxy-p-benzoquinone
Vzorec: C₈H₈O₄
MW: 168,15 g/mol
MDL Number: MFCD00016368
CAS číslo: 530-55-2
EINECS: 208-484-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR