3-Acetylcoumarin ≥98%
Dodavatel: Thermo Scientific
Synonyms:
3-Acetyl-2H-chromen-2-one
A12532.06
A12532.14
A12532.06EA
1520
CZK
A12532.06
A12532.14
3-Acetylcoumarin ≥98%
3-Acetylcoumarin
Purity: 98+%
CAS No.: 3949-36-8
Molecular Formula: C11H8O3
Formula Weight: 188.18
Melting Point: 119-124°
Beilstein Reference: 143498
Vzorec:
C₁₁H₈O₃ MW: 188,18 g/mol Bod tání: 119…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006853 CAS číslo: 3949-36-8 EINECS: 223-541-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...