Tisk…
2,2,6,6-Tetramethylpiperidinooxy (TEMPO), solid
Dodavatel: MP Biomedicals
Synonyms: TEMPO
0215268101 0215268105 0215268125
ICNA0215268101EA 1500 CZK
ICNA0215268101 ICNA0215268105 ICNA0215268125
2,2,6,6-Tetramethylpiperidinooxy (TEMPO), solid
2,2,6,6-Tetramethylpiperidinooxy (TEMPO)
TEMPO is a catalytic oxidant in the iodobenzene diacetae oxidation of nerol to neral. TEMPO has been utilised in the catalytic oxidation of primary alcohols to aldehydes.
TEMPO is a catalytic oxidant in the iodobenzene diacetae oxidation of nerol to neral. TEMPO has been utilised in the catalytic oxidation of primary alcohols to aldehydes.
Soluble in ethanol (50 mg/ml - clear, dark orange solution).
Vzorec: C₉H₁₈NO
MW: 156.25
MDL Number: MFCD00009599
CAS číslo: 2564-83-2
EINECS: 219-888-8