4-Amino-2,2,6,6-tetramethylpiperidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
115738-25G
115738-5G
SIAL115738-25GEA
8420
CZK
SIAL115738-25G
SIAL115738-5G
4-Amino-2,2,6,6-tetramethylpiperidine, Sigma-Aldrich®
4-Amino-2,2,6,6-tetramethylpiperidine
Vzorec:
C₉H₂₀N₂ MW: 156,27 g/mol |
MDL Number:
MFCD00005984 CAS číslo: 36768-62-4 EINECS: 253-197-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...