4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-Acetamido-2,2,6,6-Tetramethyl-N-Piperidinyloxy
M-Clarity™ quality level = MQ200
00375-1G
00375-5G
SIAL00375-1GEA
4280
CZK
SIAL00375-1G
SIAL00375-5G
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO), Sigma-Aldrich®
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO)
Vzorec:
C₁₁H₂₁N₂O₂ MW: 213,3 g/mol |
MDL Number:
MFCD00043593 CAS číslo: 14691-89-5 EINECS: 423-840-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...