Tisk…

2,2,6,6-Tetramethyl-4-piperidone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Triacetonamine, 2,2,6,6-Tetramethylpiperidin-4-one

M-Clarity™ quality level = MQ100

459119-100G 459119-500G
SIAL459119-100GEA 3450 CZK
SIAL459119-100G SIAL459119-500G
2,2,6,6-Tetramethyl-4-piperidone, Sigma-Aldrich®
2,2,6,6-Tetramethyl-4-piperidone
Vzorec: C₉H₁₇NO
MW: 155,24 g/mol
MDL Number: MFCD00149393
CAS číslo: 826-36-8
EINECS: 212-554-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR