Tisk…

4-Amino-1-benzylpiperidine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Amino-N-benzylpiperidine

187660050 187660250 187661000
ACRO187660050EA 1200 CZK
ACRO187660050 ACRO187660250 ACRO187661000
4-Amino-1-benzylpiperidine 98%
4-Amino-1-benzylpiperidine
Vzorec: C₁₂H₁₈N₂
MW: 190,29 g/mol
Bod varu: 152 °C (9,7 mmHg)
Hustota: 0,93 g/cm³
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006504
CAS číslo: 50541-93-0
EINECS: 256-620-9
UN: 2735
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5410 to 1.5430 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %
Sulfite (SO3) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Free chlorine ≤1 ppm (as Cl)
Ammonium (NH4) ≤3 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (as Pb) ≤1 ppm
Extractable organic subst ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR