1-Phenyl-3-pyrazolidone ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A12089.0B
A12089.18
A12089.30
A12089.0BEA
14210
CZK
A12089.0B
A12089.18
A12089.30
1-Phenyl-3-pyrazolidone ≥97%
1-Phenyl-3-pyrazolidone
Purity: 97%
CAS No.: 92-43-3
Molecular Formula: C9H10N2O
Formula Weight: 162.19
Melting Point: 120-123°
Beilstein Reference: 131856
Merck Reference: 14,7309
UN No.: 2811
MDL No.: MFCD00003094
Vzorec:
C₉H₁₀N₂O Bod tání: 120…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003094 CAS číslo: 92-43-3 EINECS: 202-155-1 UN: 2811 ADR: 6.1,III Merck Index: 14,7309 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...