Tisk…

Phenylbutazone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Butyl-1,2-diphenylpyrazolidine-3,5-dione

M-Clarity™ quality level = MQ200

P8386-100G P8386-25G P8386-500G
SIALP8386-100GEA 5360 CZK
SIALP8386-100G SIALP8386-25G SIALP8386-500G
Phenylbutazone, Sigma-Aldrich®
Phenylbutazone
Vzorec: C₁₉H₂₀N₂O₂
MDL Number: MFCD00005500
CAS číslo: 50-33-9
EINECS: 200-029-0
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR