Tisk…

Phenylbutazone

Dodavatel: Cayman Chemical

70400-1 70400-50
CAYM70400-1EA 613 CZK
CAYM70400-1 CAYM70400-50
Phenylbutazone
Phenylbutazone
Vzorec: C₁₉H₂₀N₂O₂
MDL Number: MFCD00005500
CAS číslo: 50-33-9
EINECS: 200-029-0
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR