Tisk…

Norbornene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Norbornylene

M-Clarity™ quality level = MQ100

N32407-100G N32407-500G
SIALN32407-100GEA 1400 CZK
SIALN32407-100G SIALN32407-500G
Norbornene, Sigma-Aldrich®
Norbornene
Vzorec: C₇H₁₀
MW: 94,16 g/mol
MDL Number: MFCD00082304
CAS číslo: 498-66-8
EINECS: 207-866-0
UN: 1325
ADR: 4.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR