Norbornene ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Norbornylene
L11103.22
L11103.36
L11103.22EA
934
CZK
L11103.22
L11103.36
Norbornene ≥99%
Norbornene
Purity: 99%
CAS No.: 498-66-8
Molecular Formula: C7H10
Formula Weight: 94.16
Flash Point: -15°(5°F)
Odour: Pungent
Boiling Point: 95-96°
Beilstein Reference: 1098544
UN No.: 1325
MDL No.: MFCD00082304
Vzorec:
C₇H₁₀ MW: 94,16 g/mol Bod varu: 95…96 °C Bod tání: 44…47 °C Hustota: 0,955 Bod vzplanutí: –15 °C (5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00082304 CAS číslo: 498-66-8 EINECS: 207-866-0 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...