Tisk…

4-Aminobutyraldehyde diethyl acetal, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

A44150-100G A44150-25G A44150-5G
SIALA44150-100GEA 11820 CZK
SIALA44150-100G SIALA44150-25G SIALA44150-5G
4-Aminobutyraldehyde diethyl acetal, Sigma-Aldrich®
4-Aminobutyraldehyde diethyl acetal
Vzorec: C₈H₁₉NO₂
MDL Number: MFCD00008227
CAS číslo: 6346-09-4
EINECS: 228-753-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR