Tisk…

2,5-Piperazinedione, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Glycine anhydride, Piperazine-2,5-dione

M-Clarity™ quality level = MQ200

G7251-100G G7251-1KG G7251-25G
SIALG7251-100GEA 10320 CZK
SIALG7251-100G SIALG7251-1KG SIALG7251-25G
2,5-Piperazinedione, Sigma-Aldrich®
2,5-Piperazinedione
Vzorec: C₄H₆N₂O₂
MW: 114,1 g/mol
MDL Number: MFCD00006009
CAS číslo: 106-57-0
EINECS: 203-411-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR