2-(Methylthio)ethanol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W400408-100G
W400408-1KG
W400408-SAMPLE
SIALW400408-100GEA
15440
CZK
SIALW400408-100G
SIALW400408-1KG
SIALW400408-SAMPLE
2-(Methylthio)ethanol, Sigma-Aldrich®
2-(Methylthio)ethanol
Vzorec:
C₃H₈OS MW: 92.16 |
MDL Number:
MFCD00002908 CAS číslo: 5271-38-5 EINECS: 226-090-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...