Calcium nitrate hydrate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
202967-10G
202967-50G
SIAL202967-10GEA
2620
CZK
SIAL202967-10G
SIAL202967-50G
Calcium nitrate hydrate, Sigma-Aldrich®
Calcium nitrate hydrate
Vzorec:
Ca(NO₃)₂·H₂O MW: 182,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149604 CAS číslo: 35054-52-5 EINECS: 233-332-1 UN: 1454 ADR: 5.1,III Merck Index: 13,01687 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...