Tisk…

Methoxyhydroquinone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

176893-25G 176893-5G
SIAL176893-25GEA 7470 CZK
SIAL176893-25G SIAL176893-5G
Methoxyhydroquinone, Sigma-Aldrich®
Methoxyhydroquinone
Vzorec: C₇H₈O₃
MDL Number: MFCD00013971
CAS číslo: 824-46-4
EINECS: 212-530-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR