Tisk…
sec-Butyllithium 1.3 M in cyclohexane (92% )(v/v)/hexane (mixture of isomers) (8% (v/v)), AcroSeal®
Dodavatel: Thermo Fisher Scientific
Nebezpečí
Synonyms: s-Butyllithium
187541000 187548000
187541000.EA 2170 CZK
187541000. 187548000.
sec-Butyllithium 1.3 M in cyclohexane (92% )(v/v)/hexane (mixture of isomers) (8% (v/v)), AcroSeal®
sec-Butyllithium
Vzorec: C₄H₉Li
MW: 64.06 g/mol
Hustota: 0.75 g/cm³
Bod vzplanutí: –17 °C
Storage Temperature: Freezer
MDL Number: MFCD00009323
CAS číslo: 598-30-1
EINECS: 209-927-7
UN: 3394
ADR: 4.2 I
Objednat
Výsledky specifikačního testu
Appearance Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
Titration Active base 1.23 to 1.46 M
UV at 500 nm A: ≤0.02
Gradient analysis pass test