Tisk…

L(+)-Ornithine dihydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (S)-2,5-Diaminovaleric acid dihydrochloride, H-Orn-OH.2HCl, (S)-2,5-Diaminopentanoic acid dihydrochloride, L-Ornithine dihydrochloride

M-Clarity™ quality level = MQ100

75440-100G 75440-25G
SIAL75440-100GEA 36400 CZK
SIAL75440-100G SIAL75440-25G
L(+)-Ornithine dihydrochloride, Sigma-Aldrich®
L(+)-Ornithine dihydrochloride
Vzorec: C₅H₁₄Cl₂N₂O₂
MW: 205,08 g/mol
MDL Number: MFCD00066571
CAS číslo: 6211-16-1
EINECS: 228-277-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR