L(+)-Ornithine hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
H-Orn-OH Hydrochloride, H-Orn-OH·HCl
M-Clarity™ quality level = MQ200
O4386-100G
O4386-25G
SIALO4386-100GEA
14810
CZK
SIALO4386-100G
SIALO4386-25G
L(+)-Ornithine hydrochloride, Sigma-Aldrich®
L(+)-Ornithine hydrochloride
Vzorec:
C₅H₁₂N₂O₂·HCl MW: 168.62 |
MDL Number:
MFCD00064562 CAS číslo: 3184-13-2 EINECS: 221-678-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...