Sodium-tert-butanolate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate
M-Clarity™ quality level = MQ200
359270-100G
359270-500G
359270-5G
SIAL359270-100GEA
1910
CZK
SIAL359270-100G
SIAL359270-500G
SIAL359270-5G
Sodium-tert-butanolate, Sigma-Aldrich®
Sodium-tert-butanolate
Vzorec:
C₄H₉NaO MW: 96.1 |
MDL Number:
MFCD00040575 CAS číslo: 865-48-5 EINECS: 212-741-9 UN: 3206 ADR: 4.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...