Tisk…

2-Methylpyrimidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

686956-1G 686956-250MG
SIAL686956-1GEA 13390 CZK
SIAL686956-1G SIAL686956-250MG
2-Methylpyrimidine, Sigma-Aldrich®
2-Methylpyrimidine
Vzorec: C₅H₆N₂
MW: 94,12 g/mol
MDL Number: MFCD00128182
CAS číslo: 5053-43-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR