Tisk…

Aluminium tri-tert-butanolate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Aluminium tert-butoxide, Aluminumtert-butoxide

M-Clarity™ quality level = MQ100

235849-10G 235849-50G
SIAL235849-10GEA 2260 CZK
SIAL235849-10G SIAL235849-50G
Aluminium tri-tert-butanolate, Sigma-Aldrich®
Aluminium tri-tert-butanolate
Vzorec: C₁₂H₂₇AlO₃
MW: 246.33
MDL Number: MFCD00008801
CAS číslo: 556-91-2
EINECS: 209-146-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR