Trilithium citrate hydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
213209-100G
213209-500G
SIAL213209-100GEA
0
CZK
SIAL213209-100G
SIAL213209-500G
Trilithium citrate hydrate, Sigma-Aldrich®
Trilithium citrate hydrate
Vzorec:
C₆H₅Li₃O₇·H₂O MW: 227,94 g/mol |
CAS číslo:
313222-91-2 EINECS: 213-045-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...