Tisk…

Elaidic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (E)-Octadec-9-enoic acid

E4637-1G E4637-5G
SIALE4637-1GEA 7730 CZK
SIALE4637-1G SIALE4637-5G
Elaidic acid, Sigma-Aldrich®
Elaidic acid
Vzorec: C₁₈H₃₄O₂
MW: 282,47 g/mol
MDL Number: MFCD00063954
CAS číslo: 112-79-8
EINECS: 204-006-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR