Tisk…

Elaidic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A14832.06 A14832.14 A14832.03 AB113010.0025
A14832.06EA 4810 CZK
A14832.06 A14832.14 A14832.03 ABCRAB113010.0025
Elaidic acid ≥98%
Elaidic acid

Purity: 98% CAS No.: 112-79-8 Molecular Formula: C18H34O2 Formula Weight: 282.47 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 41-45° Boiling Point: 288°/100mm Beilstein Reference: 1726543 Merck Reference: 14,3533 MDL No.: MFCD00063954

Vzorec: C₁₈H₃₄O₂
Bod varu: 288 °C (100 mmHg)
Bod tání: 41…45 °C
Bod vzplanutí: >110 °C (230 °F)
MDL Number: MFCD00063954
CAS číslo: 112-79-8
EINECS: 204-006-6
Merck Index: 14,3533

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR