Ethyl mercaptoacetate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Ethyl 2-mercaptoacetate, Ethyl thioglycolate
A14321.06
A14321.22
A14321.36
A14321.06EA
745
CZK
A14321.06
A14321.22
A14321.36
Ethyl mercaptoacetate ≥98%
Ethyl mercaptoacetate
Purity: 98+%
CAS No.: 623-51-8
Molecular Formula: C4H8O2S
Formula Weight: 120.17
Flash Point: 48°(118°F)
Refractive Index: 1.4580
Odour: Stench
Melting Point: -80°
Boiling Point: 156-159°
Beilstein Reference: 605581
UN No.: 1228
MDL No.: MFCD00004874
Vzorec:
C₄H₈O₂S Bod varu: 156…159 °C Bod tání: –80 °C Hustota: 1,096 Bod vzplanutí: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004874 CAS číslo: 623-51-8 EINECS: 210-800-3 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...