2-Chloro-5-fluoropyrimidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
651753-1G
651753-250MG
SIAL651753-1GEA
9900
CZK
SIAL651753-1G
SIAL651753-250MG
2-Chloro-5-fluoropyrimidine, Sigma-Aldrich®
2-Chloro-5-fluoropyrimidine
Vzorec:
C₄H₂ClFN₂ MW: 132,52 g/mol |
MDL Number:
MFCD03788197 CAS číslo: 62802-42-0 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...