4,6-Dichloro-2-methylpyrimidine ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H50493.03
H50493.06
H50493.03EA
1800
CZK
H50493.03
H50493.06
4,6-Dichloro-2-methylpyrimidine ≥97%
4,6-Dichloro-2-methylpyrimidine
Purity: 97%
CAS No.: 1780-26-3
Molecular Formula: C5H4Cl2N2
Formula Weight: 163.01
Melting Point: 41-45°
UN No.: 1759
MDL No.: MFCD00090472
Vzorec:
C₅H₄Cl₂N₂ Bod tání: 41…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090472 CAS číslo: 1780-26-3 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...