Tisk…
Pyridoxal hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl
M-Clarity™ quality level = MQ200
271748-1G 271748-25G 271748-5G
SIAL271748-1GEA 1750 CZK
SIAL271748-1G SIAL271748-25G SIAL271748-5G
Pyridoxal hydrochloride, Sigma-Aldrich®
Pyridoxal hydrochloride
Vzorec: C₈H₁₀ClNO₃
MW: 203.62
MDL Number: MFCD00012809
CAS číslo: 65-22-5
EINECS: 200-602-5