Tetrahydrofuran unstabilised for HPLC
Dodavatel: SCHARLAB
|
Nebezpečí
|
Synonyms:
Butylene oxide, THF, Diethylene oxide
TE02251000
TE02252500
SCARTE02251000EA
0
CZK
SCARTE02251000
SCARTE02252500
Tetrahydrofuran unstabilised for HPLC
Tetrahydrofuran
Vzorec:
C₄H₈O Bod varu: 66 °C (1013 hPa) Hustota: 0,888 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005356 CAS číslo: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3,II REACH: 05-2114615188-48 Merck Index: 12,09351 |