Tisk…

Kyselina mravenčí 95% ≥95%, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Aminic Acid, Methanoic acid

M-Clarity™ quality level = MQ300

W248703-10KG-K W248703-1KG-K W248703-25KG-K W248703-5KG-K
SIALW248703-10KG-KEA 13940 CZK
SIALW248703-10KG-K SIALW248703-1KG-K SIALW248703-25KG-K SIALW248703-5KG-K
Kyselina mravenčí 95% ≥95%, Sigma-Aldrich®
Kyselina mravenčí
Vzorec: HCO₂H
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS číslo: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR