Dextran, M.W. ~ 70,000 (from Leuconostoc mesenteroides), Sigma-Aldrich®
Dodavatel: Merck
D8821-500G
D8821-100G
D8821-10G
D8821-50G
SIALD8821-500GEA
78070
CZK
SIALD8821-500G
SIALD8821-100G
SIALD8821-10G
SIALD8821-50G
Dextran, M.W. ~ 70,000 (from Leuconostoc mesenteroides), Sigma-Aldrich®
Dextran
Vzorec:
(C₆H₁₀O₅)n |
MDL Number:
MFCD00130935 CAS číslo: 9004-54-0 EINECS: 232-677-5 |