Tisk…

Benzyltrichlorosilane

Dodavatel: abcr

AB110825.0100 AB110825.0025
ABCRAB110825.0100EA 0 CZK
ABCRAB110825.0100 ABCRAB110825.0025
Benzyltrichlorosilane
Benzyltrichlorosilane
Vzorec: C₇H₇Cl₃Si
CAS číslo: 770-10-5
EINECS: 212-219-0
UN: 2987
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR