Decamethylcyclopentasiloxane
Dodavatel: abcr
Synonyms:
Cyclopentasiloxane, 2,2,4,4,6,6,8,8,10,10-Decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane, Dow Corning 245® fluid, Volasil 245, Silicone fluid DC 245
AB111012.2000
AB111012.0100
ABCRAB111012.2000EA
0
CZK
ABCRAB111012.2000
ABCRAB111012.0100
Decamethylcyclopentasiloxane
Decamethylcyclopentasiloxane
Vzorec:
C₁₀H₃₀O₅Si₅ MW: 370.77 Bod varu: 90 Bod tání: –44 °C Hustota: 0.958 Bod vzplanutí: 73 °C (ot. pohárek) Storage Temperature: Ambient |
MDL Number:
MFCD00046966 CAS číslo: 541-02-6 EINECS: 208-764-9 Merck Index: 12,02905 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...