Tisk…
Vario couplings
Dodavatel: Bohlender
F778-08 F778-16 D681-08 D681-24 D681-32 D681-16 D681-40 D681-48
BOHLF778-08EA 1200 CZK
BOHLF778-08 BOHLF778-16 BOHLD681-08 BOHLD681-24 BOHLD681-32 BOHLD681-16 BOHLD681-40 BOHLD681-48
Vario couplings
Fitinky a konektory
Vario 2-part coupling to connect elastic tubing with non-elastic tubing made of PTFE, glass, metal or other plastics.
  • The elastic tubing is pushed on a cone and fixed on the coupling by means of a connecting nut
  • These couplings are available in two different versions, according to male and female threads to connect