1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H51065.MD
H51065.03
H51065.06
H51065.MDEA
3190
CZK
H51065.MD
H51065.03
H51065.06
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) 95%
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane)
Purity: 95%
CAS No.: 33941-15-0
Molecular Formula: C12H25NO5
Formula Weight: 263.33
Melting Point: 46-49°
Beilstein Reference: 1074079
Vzorec:
C₁₂H₂₅NO₅ Bod tání: 46…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075466 CAS číslo: 33941-15-0 EINECS: 251-751-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...