Dicyklohexylkarbodiimid 1 mol/l in dichloromethane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,3-Dicyclohexylcarbodiimide, DCC, N,N'-Dicyclohexylcarbodiimide
M-Clarity™ quality level = MQ200
379115-100ML
379115-800ML
SIAL379115-100MLEA
1850
CZK
SIAL379115-100ML
SIAL379115-800ML
Dicyklohexylkarbodiimid 1 mol/l in dichloromethane, Sigma-Aldrich®
Dicyklohexylkarbodiimid
Vzorec:
C₁₃H₂₂N₂ |
MDL Number:
MFCD00011659 CAS číslo: 538-75-0 EINECS: 208-704-1 Merck Index: 13,03123 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...