4,4'-Bis(dimethylamino)diphenylmethane ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98+%
CAS No.: 101-61-1
Molecular Formula: C17H22N2
Formula Weight: 254.38
Flash Point: 178°(352°F)
Melting Point: 85-88°
Boiling Point: 155-157°/0.1mm
Beilstein Reference: 479956
Merck Reference: 14,6176
UN No.: 3077
MDL No.: MFCD00008317
Vzorec:
C₁₇H₂₂N₂ Bod varu: 155…157 °C (0,1 mmHg) Bod tání: 85…88 °C Bod vzplanutí: 178 °C (352 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008317 CAS číslo: 101-61-1 EINECS: 202-959-2 ADR: 9,III Merck Index: 14,6176 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...