Tisk…

Benzyl carbazate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate

499781-25G 499781-5G
SIAL499781-25GEA 7610 CZK
SIAL499781-25G SIAL499781-5G
Benzyl carbazate, Sigma-Aldrich®
Benzyl carbazate
Vzorec: C₈H₁₀N₂O₂
MW: 166,18 g/mol
MDL Number: MFCD00041890
CAS číslo: 5331-43-1
EINECS: 226-230-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR