Benzyl carbazate ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate
A13005.06
A13005.14
A13005.22
A13005.06EA
1820
CZK
A13005.06
A13005.14
A13005.22
Benzyl carbazate ≥98%
Benzyl carbazate
Purity: 98+%
CAS No.: 5331-43-1
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Melting Point: 66-70°
Beilstein Reference: 1952982
Vzorec:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Bod tání: 66…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041890 CAS číslo: 5331-43-1 EINECS: 226-230-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...