Tisk…
Potassium thioacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Potassium ethanethioate
M-Clarity™ quality level = MQ100
241776-100G 241776-25G
SIAL241776-100GEA 5560 CZK
SIAL241776-100G SIAL241776-25G
Potassium thioacetate, Sigma-Aldrich®
Potassium thioacetate
Vzorec: C₂H₃KOS
MDL Number: MFCD00137704
CAS číslo: 10387-40-3
EINECS: 233-848-7