S-Ethyl thioacetate ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A10680.06
A10680.14
A10680.06EA
838
CZK
A10680.06
A10680.14
S-Ethyl thioacetate ≥98%
S-Ethyl thioacetate
Purity: 98+%
CAS No.: 625-60-5
Molecular Formula: C4H8OS
Formula Weight: 104.18
Flash Point: 20°(68°F)
Refractive Index: 1.4600
Odour: Stench
Boiling Point: 116-117°
Solubility: Insoluble in water
Beilstein Reference: 1737643
UN No.: 1993
MDL No.: MFCD00015178
Vzorec:
C₄H₈OS MW: 104,17 g/mol Bod varu: 116…117 °C Hustota: 0,980 Bod vzplanutí: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015178 CAS číslo: 625-60-5 EINECS: 210-904-9 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...