Formamidine acetate ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A11158.22
A11158.36
A11158.22EA
1770
CZK
A11158.22
A11158.36
Formamidine acetate ≥99%
Formamidine acetate
Purity: 99%
CAS No.: 3473-63-0
Molecular Formula: C3H8N2O2
Formula Weight: 104.11
pH: 8
Melting Point: ca 158° dec.
Beilstein Reference: 3563488
MDL No.: MFCD00012866
Vzorec:
C₃H₈N₂O₂ MW: 104.11 Bod tání: ∼ 158 °C (rozkl.) Hustota: 1.124 Storage Temperature: Ambient |
MDL Number:
MFCD00012866 CAS číslo: 3473-63-0 EINECS: 222-442-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...